Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-trans-4-fluoro-L-proline methyl ester, 97%

$119.65 - $376.69

Chemical Identifiers

Chemical Name or Material N-Boc-trans-4-fluoro-L-proline methyl ester
CAS 203866-18-6
Molecular Formula C11H18FNO4
Formula Weight 247.27
MDL Number MFCD07367993
Synonym (2S,4R)-4-Fluoropyrrolidine-1,2-dicarboxylic acid 1-tert-butyl 2-methyl diester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2774703

  SDS
alfa aesar™
H2774703
1g Each for $376.69

AAH27747MD

  SDS
alfa aesar™
H27747MD
250mg Each for $119.65
Description & Specifications

Specifications

Formula Weight 247.27
Quantity 1g
Molecular Formula C11H18FNO4
Assay 97%
MDL Number MFCD07367993
CAS 203866-18-6
Recommended Storage 2° to 8°C
Synonym (2S,4R)-4-Fluoropyrrolidine-1,2-dicarboxylic acid 1-tert-butyl 2-methyl diester
Melting Point 36° to 39°C
Chemical Name or Material N-Boc-trans-4-fluoro-L-proline methyl ester

Applications
N-Boc-trans-4-fluoro-L-proline methyl ester is used to prepare (S)-2-[N-(N′-benzyl-(S)-4-fluoro-L-prolyl)amino]benzophenone by reacting with of 2-aminobenzophenone in the presence of potassium tert -butoxide.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Safety and Handling

2° to 8°C