Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-trans-4-fluoro-L-proline, 98%

$128.38 - $323.24

Chemical Identifiers

Chemical Name or Material N-Boc-trans-4-fluoro-L-proline
CAS 203866-14-2
Molecular Formula C10H16FNO4
Formula Weight 233.24
MDL Number MFCD06796085
Synonym (2S,4R)-4-Fluoropyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2772103

 
alfa aesar™
H2772103
1g Each for $323.24

AAH27721MD

 
alfa aesar™
H27721MD
250mg Each for $128.38
Description & Specifications

Specifications

Formula Weight 233.24
Chemical Name or Material N-Boc-trans-4-fluoro-L-proline
Assay 98%
Molecular Formula C10H16FNO4
Melting Point 115° to 117°C
MDL Number MFCD06796085
CAS 203866-14-2
Quantity 1g
Synonym (2S,4R)-4-Fluoropyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C