missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Boc-thiourea, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6359903

 View more versions of this product

Catalog No. AAH6359903


Description

Description

N-Boc-thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

142°C (decomposition)
268551-65-1
C6H12N2O2S
n-boc-thiourea, n-tert-butoxycarbonylthiourea, n-boc thiourea, mono-boc-thiourea, acmc-20ak5u, n-tert-butoxycarbonyl-thiourea, n-tert.butoxycarbonyl thiourea, 1-tert-butoxycarbonyl thiourea, tert-butyl carbamothioylcarbamate
CC(C)(C)OC(=O)NC(N)=S
176.23
176
1 g
N-Boc-thiourea
White
MFCD04039987
JKXQTODLIZBUDE-UHFFFAOYSA-N
tert-butyl N-carbamothioylcarbamate
4095660
97%
Powder

Chemical Identifiers

268551-65-1
176.23
JKXQTODLIZBUDE-UHFFFAOYSA-N
4095660
CC(C)(C)OC(=O)NC(N)=S
C6H12N2O2S
MFCD04039987
n-boc-thiourea, n-tert-butoxycarbonylthiourea, n-boc thiourea, mono-boc-thiourea, acmc-20ak5u, n-tert-butoxycarbonyl-thiourea, n-tert.butoxycarbonyl thiourea, 1-tert-butoxycarbonyl thiourea, tert-butyl carbamothioylcarbamate
tert-butyl N-carbamothioylcarbamate
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c

H317-H334

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only