missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-thiourea, 97%

$41.46 - $479.41

Chemical Identifiers

CAS 268551-65-1
Molecular Formula C6H12N2O2S
Molecular Weight (g/mol) 176.234
MDL Number MFCD04039987
InChI Key JKXQTODLIZBUDE-UHFFFAOYSA-N
Synonym 1-tert-butoxycarbonyl thiourea, acmc-20ak5u, mono-boc-thiourea, n-boc thiourea, n-boc-thiourea, n-tert-butoxycarbonyl-thiourea, n-tert-butoxycarbonylthiourea, n-tert.butoxycarbonyl thiourea, tert-butyl carbamothioylcarbamate
PubChem CID 4095660
IUPAC Name tert-butyl N-carbamothioylcarbamate
SMILES CC(C)(C)OC(=O)NC(=S)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63599MD View Documents Alfa Aesar™
H63599MD
250mg Each for $41.46
 
AAH6359903 View Documents Alfa Aesar™
H6359903
1g Each for $123.85
 
AAH6359906 View Documents Alfa Aesar™
H6359906
5g Each for $479.41
 
Description

Description

Applications
N-Boc-thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

268551-65-1
176.234
JKXQTODLIZBUDE-UHFFFAOYSA-N
4095660
CC(C)(C)OC(=O)NC(=S)N
C6H12N2O2S
MFCD04039987
1-tert-butoxycarbonyl thiourea, acmc-20ak5u, mono-boc-thiourea, n-boc thiourea, n-boc-thiourea, n-tert-butoxycarbonyl-thiourea, n-tert-butoxycarbonylthiourea, n-tert.butoxycarbonyl thiourea, tert-butyl carbamothioylcarbamate
tert-butyl N-carbamothioylcarbamate

Specifications

White
250mg
C6H12N2O2S
1-tert-butoxycarbonyl thiourea, acmc-20ak5u, mono-boc-thiourea, n-boc thiourea, n-boc-thiourea, n-tert-butoxycarbonyl-thiourea, n-tert-butoxycarbonylthiourea, n-tert.butoxycarbonyl thiourea, tert-butyl carbamothioylcarbamate
CC(C)(C)OC(=O)NC(=S)N
176.234
176
97%
142°C (decomposition)
268551-65-1
MFCD04039987
JKXQTODLIZBUDE-UHFFFAOYSA-N
tert-butyl N-carbamothioylcarbamate
4095660
Powder
N-Boc-thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c

H317-H334

TSCA : No

Recommended Storage : Ambient temperatures