missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-thiourea, 97%

Manufacturer:  Alfa Aesar™ H63599MD

 View more versions of this product

Catalog No. AAH63599MD



Description

Description

Applications
N-Boc-thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

N-Boc-thiourea
142°C (decomposition)
268551-65-1
MFCD04039987
JKXQTODLIZBUDE-UHFFFAOYSA-N
tert-butyl N-carbamothioylcarbamate
4095660
Powder
White
250mg
C6H12N2O2S
n-boc-thiourea, n-tert-butoxycarbonylthiourea, n-boc thiourea, mono-boc-thiourea, acmc-20ak5u, n-boc-thiourea, n-tert-butoxycarbonyl-thiourea, n-tert.butoxycarbonyl thiourea, 1-tert-butoxycarbonyl thiourea, tert-butyl carbamothioylcarbamate
CC(C)(C)OC(=O)NC(=S)N
176.234
176
97%

Chemical Identifiers

268551-65-1
176.234
JKXQTODLIZBUDE-UHFFFAOYSA-N
4095660
CC(C)(C)OC(=O)NC(=S)N
C6H12N2O2S
MFCD04039987
n-boc-thiourea, n-tert-butoxycarbonylthiourea, n-boc thiourea, mono-boc-thiourea, acmc-20ak5u, n-boc-thiourea, n-tert-butoxycarbonyl-thiourea, n-tert.butoxycarbonyl thiourea, 1-tert-butoxycarbonyl thiourea, tert-butyl carbamothioylcarbamate
tert-butyl N-carbamothioylcarbamate
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents