missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-sarcosine methyl ester, 97%

Manufacturer:  Alfa Aesar™ H6230406

 View more versions of this product

Catalog No. AAH6230406



Description

Description

Applications
N-Boc-sarcosine methyl ester is involved in the preparation of N-[(R, R)-(E)-1-arylmethyl-3-(2-oxo-azepan-3-yl) carbamoyl] allyl-N-methyl-3, 5-bis(trifluoromethyl) benzamides.

Notes
Incompatible with oxidizing agents. Heat sensitive.
Specifications

Specifications

N-Boc-sarcosine methyl ester
C9H17NO4
MFCD04973115
LTBDTYJYKRLTRN-UHFFFAOYSA-N
methyl 2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetate
14018930
Liquid
Yellow
42492-57-9
(CH3)3COCON(CH3)CH2CO2CH3
boc-gly-ome
CC(C)(C)OC(=O)N(C)CC(=O)OC
203.238
203.24
97%
5g

Chemical Identifiers

42492-57-9
203.238
LTBDTYJYKRLTRN-UHFFFAOYSA-N
14018930
CC(C)(C)OC(=O)N(C)CC(=O)OC
C9H17NO4
MFCD04973115
boc-gly-ome
methyl 2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetate
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

Documents