Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-S-tert-butyl-L-cysteine, 98%

Chemical Identifiers

Chemical Name or Material N-Boc-S-tert-butyl-L-cysteine
CAS 56976-06-8
Formula Weight 277.38
MDL Number MFCD00076918
Synonym Boc-Cys(tBu)-OH; (R)-2-(Boc-amino)-3-(tert-butylthio)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6339906

  SDS
alfa aesar™
H6339906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6339914

  SDS
alfa aesar™
H6339914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6339922

  SDS
alfa aesar™
H6339922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 277.38
Physical Form Solid
Assay 98%
Chemical Name or Material N-Boc-S-tert-butyl-L-cysteine
MDL Number MFCD00076918
Synonym Boc-Cys(tBu)-OH; (R)-2-(Boc-amino)-3-(tert-butylthio)propionic acid
Quantity 5g
Storage Note 1 -30° to -10°C
CAS 56976-06-8
Safety and Handling

-30° to -10°C