missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-S-tert-butyl-L-cysteine, 98%

Manufacturer:  Alfa Aesar™ H6339922

 View more versions of this product

Catalog No. AAH6339922



Specifications

Specifications

N-Boc-S-tert-butyl-L-cysteine
C12H23NO4S
boc-cys tbu-oh, boc-s-tert-butyl-l-cysteine, boc-s-t-butyl-l-cysteine, boc-l-cys tbu-oh, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-1,1-dimethylethyl, 2r-2-tert-butoxycarbonyl amino-3-tert-butylsulfanyl propanoic acid, ambotzbaa1082, boc-s-t-butyl-l-cys, boc-s-tert-butyl-l-cys, n-boc-s-tert-butyl-l-cysteine
CC(C)(C)OC(=O)NC(CSC(C)(C)C)C(=O)O
277.379
277.38
98%
56976-06-8
MFCD00076918
OGARKMDCQCLMCS-QMMMGPOBSA-N
(2R)-3-tert-butylsulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
11288930
Solid
100g

Chemical Identifiers

56976-06-8
277.379
OGARKMDCQCLMCS-QMMMGPOBSA-N
11288930
CC(C)(C)OC(=O)NC(CSC(C)(C)C)C(=O)O
C12H23NO4S
MFCD00076918
boc-cys tbu-oh, boc-s-tert-butyl-l-cysteine, boc-s-t-butyl-l-cysteine, boc-l-cys tbu-oh, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-1,1-dimethylethyl, 2r-2-tert-butoxycarbonyl amino-3-tert-butylsulfanyl propanoic acid, ambotzbaa1082, boc-s-t-butyl-l-cys, boc-s-tert-butyl-l-cys, n-boc-s-tert-butyl-l-cysteine
(2R)-3-tert-butylsulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

SDS Documents