missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-S-ethyl-L-cysteine

$101.28 - $438.69

Chemical Identifiers

CAS 16947-82-3
Molecular Formula C10H19NO4S
Formula Weight 249.33
MDL Number MFCD00076919
Synonym N-(tert-butoxycarbonyl)-S-ethyl-L-cysteine
Chemical Name or Material N-Boc-S-ethyl-L-cysteine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5021603 SDS Alfa Aesar™
H5021603
1g Each for $101.28
AAH5021606 SDS Alfa Aesar™
H5021606
5g Each for $438.69
Specifications Description & Specifications

Chemical Identifiers

16947-82-3
249.33
N-(tert-butoxycarbonyl)-S-ethyl-L-cysteine
C10H19NO4S
MFCD00076919
N-Boc-S-ethyl-L-cysteine

Specifications

16947-82-3
249.33
1g
N-Boc-S-ethyl-L-cysteine
C10H19NO4S
MFCD00076919
N-(tert-butoxycarbonyl)-S-ethyl-L-cysteine

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning