Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-(S)-cyclopropylalanine benzyl ester, 95%

$521.34 - $521.34

Chemical Identifiers

Chemical Name or Material N-Boc-(S)-cyclopropylalanine benzyl ester
CAS 406681-37-6
Molecular Formula C18H25NO4
Formula Weight 319.4
MDL Number MFCD08060934
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH25758MD

  SDS
alfa aesar™
H25758MD
250mg Each for $521.34
Description & Specifications

Specifications

Formula Weight 319.4
Chemical Name or Material N-Boc-(S)-cyclopropylalanine benzyl ester
MDL Number MFCD08060934
Quantity 250mg
Melting Point 52° to 54°C
Molecular Formula C18H25NO4
CAS 406681-37-6
Assay 95%