missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-S-benzyl-L-cysteine, 98%, Thermo Scientific Chemicals
$47.90 - $149.25
Chemical Identifiers
CAS | 5068-28-0 |
---|---|
Molecular Formula | C15H21NO4S |
Molecular Weight (g/mol) | 311.40 |
MDL Number | MFCD00065567 |
InChI Key | IFVORPLRHYROAA-UHFFFAOYNA-N |
Synonym | boc-s-benzyl-l-cysteine, boc-cys bzl-oh, r-3-benzylthio-2-tert-butoxycarbonyl amino propanoic acid, n-boc-s-benzyl-l-cysteine, n-tert-butoxycarbonyl-s-benzyl-l-cysteine, s-benzyl-n-tert-butoxycarbonyl-l-cysteine, 2r-3-benzylsulfanyl-2-tert-butoxycarbonyl amino propanoic acid, boc-s-benzyl-d-cys, boc-s-benzyl-l-cys, alanine, 3-benzylthio-n-carboxy-, n-tert-butyl ester, l |
PubChem CID | 2724757 |
SMILES | CC(C)(C)OC(=O)NC(CSCC1=CC=CC=C1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6373606
|
Thermo Scientific Chemicals
H6373606 |
5 g |
Each for $47.90
|
|
|||||
AAH6373614
|
Thermo Scientific Chemicals
H6373614 |
25 g |
Each for $149.25
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5068-28-0 | |
311.40 | |
IFVORPLRHYROAA-UHFFFAOYNA-N | |
2724757 |
C15H21NO4S | |
MFCD00065567 | |
boc-s-benzyl-l-cysteine, boc-cys bzl-oh, r-3-benzylthio-2-tert-butoxycarbonyl amino propanoic acid, n-boc-s-benzyl-l-cysteine, n-tert-butoxycarbonyl-s-benzyl-l-cysteine, s-benzyl-n-tert-butoxycarbonyl-l-cysteine, 2r-3-benzylsulfanyl-2-tert-butoxycarbonyl amino propanoic acid, boc-s-benzyl-d-cys, boc-s-benzyl-l-cys, alanine, 3-benzylthio-n-carboxy-, n-tert-butyl ester, l | |
CC(C)(C)OC(=O)NC(CSCC1=CC=CC=C1)C(O)=O |
Specifications
86°C to 88°C | |
C15H21NO4S | |
2000417 | |
IFVORPLRHYROAA-UHFFFAOYNA-N | |
(2R)-3-benzylsulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
2724757 | |
98% | |
Solid |
5068-28-0 | |
MFCD00065567 | |
boc-s-benzyl-l-cysteine, boc-cys bzl-oh, r-3-benzylthio-2-tert-butoxycarbonyl amino propanoic acid, n-boc-s-benzyl-l-cysteine, n-tert-butoxycarbonyl-s-benzyl-l-cysteine, s-benzyl-n-tert-butoxycarbonyl-l-cysteine, 2r-3-benzylsulfanyl-2-tert-butoxycarbonyl amino propanoic acid, boc-s-benzyl-d-cys, boc-s-benzyl-l-cys, alanine, 3-benzylthio-n-carboxy-, n-tert-butyl ester, l | |
CC(C)(C)OC(=O)NC(CSCC1=CC=CC=C1)C(O)=O | |
311.40 | |
311.4 | |
5 g | |
N-Boc-S-benzyl-L-cysteine |
Safety and Handling
EINECSNumber : 225-772-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only