missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-S-benzyl-L-cysteine, 98%

Manufacturer:  Alfa Aesar™ H6373622

 View more versions of this product

Catalog No. AAH6373622


 

Specifications

Specifications

N-Boc-S-benzyl-L-cysteine
98%
MFCD00065567
Boc-Cys(Bzl)-OH; (R)-3-Benzylthio-2-(Boc-amino)propionic acid
CC(C)(C)OC(=O)NC(CSCC1=CC=CC=C1)C(=O)O
311.396
311.4
86°C to 88°C
5068-28-0
C15H21NO4S
2000417
IFVORPLRHYROAA-LBPRGKRZSA-N
(2R)-3-benzylsulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
2724757
Solid
100g

Chemical Identifiers

5068-28-0
311.396
IFVORPLRHYROAA-LBPRGKRZSA-N
2724757
C15H21NO4S
MFCD00065567
Boc-Cys(Bzl)-OH; (R)-3-Benzylthio-2-(Boc-amino)propionic acid
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents