missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-S-(4-methoxybenzyl)-L-cysteine, 98%, Thermo Scientific™
$46.75 - $149.25
Chemical Identifiers
CAS | 18942-46-6 |
---|---|
Molecular Formula | C16H23NO5S |
Molecular Weight (g/mol) | 341.42 |
MDL Number | MFCD00038254 |
InChI Key | VRTXRNJMNFVTOM-UHFFFAOYNA-N |
Synonym | boc-cys pmeobzl-oh, boc-s-4-methoxybenzyl-l-cysteine, boc-cys 4-meobzl-oh, boc-cys mob-oh, r-2-tert-butoxycarbonyl amino-3-4-methoxybenzyl thio propanoic acid, n-tert-butoxycarbonyl-s-4-methoxybenzyl-l-cysteine, 2r-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, n-tert-butoxycarbonyl-s-p-methoxybenzyl-l-cysteine, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-4-methoxyphenyl methyl, n-1,1-dimethylethoxy carbonyl-s-4-methoxyphenyl methyl-l-cysteine |
PubChem CID | 87863 |
IUPAC Name | (2R)-3-[(4-methoxyphenyl)methylsulfanyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
SMILES | COC1=CC=C(CSCC(NC(=O)OC(C)(C)C)C(O)=O)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6380806
|
Thermo Scientific Chemicals
H6380806 |
5 g |
Each for $46.75
|
|
|||||
AAH6380814
|
Thermo Scientific Chemicals
H6380814 |
25 g |
Each for $149.25
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
18942-46-6 | |
341.42 | |
VRTXRNJMNFVTOM-UHFFFAOYNA-N | |
87863 | |
COC1=CC=C(CSCC(NC(=O)OC(C)(C)C)C(O)=O)C=C1 |
C16H23NO5S | |
MFCD00038254 | |
boc-cys pmeobzl-oh, boc-s-4-methoxybenzyl-l-cysteine, boc-cys 4-meobzl-oh, boc-cys mob-oh, r-2-tert-butoxycarbonyl amino-3-4-methoxybenzyl thio propanoic acid, n-tert-butoxycarbonyl-s-4-methoxybenzyl-l-cysteine, 2r-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, n-tert-butoxycarbonyl-s-p-methoxybenzyl-l-cysteine, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-4-methoxyphenyl methyl, n-1,1-dimethylethoxy carbonyl-s-4-methoxyphenyl methyl-l-cysteine | |
(2R)-3-[(4-methoxyphenyl)methylsulfanyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
18942-46-6 | |
MFCD00038254 | |
VRTXRNJMNFVTOM-UHFFFAOYNA-N | |
(2R)-3-[(4-methoxyphenyl)methylsulfanyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
87863 | |
98% | |
Solid |
C16H23NO5S | |
boc-cys pmeobzl-oh, boc-s-4-methoxybenzyl-l-cysteine, boc-cys 4-meobzl-oh, boc-cys mob-oh, r-2-tert-butoxycarbonyl amino-3-4-methoxybenzyl thio propanoic acid, n-tert-butoxycarbonyl-s-4-methoxybenzyl-l-cysteine, 2r-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, n-tert-butoxycarbonyl-s-p-methoxybenzyl-l-cysteine, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-4-methoxyphenyl methyl, n-1,1-dimethylethoxy carbonyl-s-4-methoxyphenyl methyl-l-cysteine | |
COC1=CC=C(CSCC(NC(=O)OC(C)(C)C)C(O)=O)C=C1 | |
341.42 | |
341.42 | |
5 g | |
N-Boc-S-(4-methoxybenzyl)-L-cysteine |
Safety and Handling
EINECSNumber : 242-695-5
TSCA : Yes
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold