missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-S-(4-methoxybenzyl)-L-cysteine, 98%

Manufacturer:  Alfa Aesar™ H6380822

 View more versions of this product

Catalog No. AAH6380822



Specifications

Specifications

N-Boc-S-(4-methoxybenzyl)-L-cysteine
C16H23NO5S
boc-cys pmeobzl-oh, boc-s-4-methoxybenzyl-l-cysteine, boc-cys 4-meobzl-oh, boc-cys mob-oh, r-2-tert-butoxycarbonyl amino-3-4-methoxybenzyl thio propanoic acid, n-tert-butoxycarbonyl-s-4-methoxybenzyl-l-cysteine, 2r-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, n-tert-butoxycarbonyl-s-p-methoxybenzyl-l-cysteine, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-4-methoxyphenyl methyl, n-1,1-dimethylethoxy carbonyl-s-4-methoxyphenyl methyl-l-cysteine
CC(C)(C)OC(=O)NC(CSCC1=CC=C(C=C1)OC)C(=O)O
341.422
341.42
98%
18942-46-6
MFCD00038254
VRTXRNJMNFVTOM-ZDUSSCGKSA-N
(2R)-3-[(4-methoxyphenyl)methylsulfanyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
87863
Solid
100g

Chemical Identifiers

18942-46-6
341.422
VRTXRNJMNFVTOM-ZDUSSCGKSA-N
87863
CC(C)(C)OC(=O)NC(CSCC1=CC=C(C=C1)OC)C(=O)O
C16H23NO5S
MFCD00038254
boc-cys pmeobzl-oh, boc-s-4-methoxybenzyl-l-cysteine, boc-cys 4-meobzl-oh, boc-cys mob-oh, r-2-tert-butoxycarbonyl amino-3-4-methoxybenzyl thio propanoic acid, n-tert-butoxycarbonyl-s-4-methoxybenzyl-l-cysteine, 2r-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, n-tert-butoxycarbonyl-s-p-methoxybenzyl-l-cysteine, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-4-methoxyphenyl methyl, n-1,1-dimethylethoxy carbonyl-s-4-methoxyphenyl methyl-l-cysteine
(2R)-3-[(4-methoxyphenyl)methylsulfanyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Safety and Handling

Safety and Handling

TSCA : TSCA

storageNote1 : 2° to 8°C

SDS Documents