Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-S-(3-nitro-2-pyridylthio)-L-cysteine, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-S-(3-nitro-2-pyridylthio)-L-cysteine
CAS 76880-29-0
Formula Weight 375.41
MDL Number MFCD00153301
Beilstein 3569570
Synonym Boc-Cys(Npys)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6621306

 
alfa aesar™
H6621306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6621303

 
alfa aesar™
H6621303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 375.41
Chemical Name or Material N-Boc-S-(3-nitro-2-pyridylthio)-L-cysteine
CAS 76880-29-0
Assay 95%
MDL Number MFCD00153301
Synonym Boc-Cys(Npys)-OH
Quantity 5g
Sensitivity Moisture Sensitive
Physical Form Powder
Beilstein 3569570
Color Pale yellow
Melting Point ≈160°C (dec.)

Applications
N-Boc-S-(3-nitro-2-pyridylthio)-L-cysteine is used as medical intermediates.

Notes
Moisture Sensitive. Store under dry inert gas. Store away from moisture. Protect from humidity and water. Incompatible with oxidizing agents.