Please login to your online account to display your discounted pricing

N-BOC-Pyrrolidine, 97%, Acros Organics


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC425820010

 View more versions of this product

Catalog No. AC425820010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 171.24
Quantity 1g
Chemical Name or Material N-BOC-Pyrrolidine
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C9H17NO2
CAS 86953-79-9

Specifications

Chemical Name or Material N-BOC-Pyrrolidine
Assay Percent Range 97%
Molecular Formula C9H17NO2
Formula Weight 171.24