missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-propargylamine, 97%

Manufacturer:  Alfa Aesar™ H5340603

 View more versions of this product

Catalog No. AAH5340603



Description

Description

Applications
N-Boc-propargylamine is used to prepare triazolobenzylidene-thiazolopyrimidines. which act as CDC25 phosphatase inhibitors. Further, it is used for the synthesis of beta-glucan polysaccharide analogs. In addition to this, it is involved in the Pauson-Khand (PK) reaction of norbornadiene and N-Boc-propargylamine to prepare 4,5-disubstituted cyclopentenones .

Solubility
Soluble in chloroform.

Notes
Moisture sensitive. Incompatible with oxidizing agents. Store in a cool place.
Specifications

Specifications

N-Boc-propargylamine
40°C to 44°C
92136-39-5
HC≡CCH2NHCO2C(CH3)3
n-boc-propargylamine, n-tert-butoxycarbonyl propargylamine, n-boc-prop-2-ynylamine, tert-butyl prop-2-yn-1-ylcarbamate, n-boc-propargyl amine, tert-butyl n-prop-2-yn-1-yl carbamate, tert-butyl prop-2-ynylcarbamate, n-boc-2-propyn-1-amine, n-boc propargyl amine, prop-2-ynyl-carbamic acid tert-butyl ester
CC(C)(C)OC(=O)NCC#C
155.197
155.2
93°C (199°F)
1g
C8H13NO2
MFCD07367245
DSPYCWLYGXGJNJ-UHFFFAOYSA-N
tert-butyl N-prop-2-ynylcarbamate
11073648
97%

Chemical Identifiers

92136-39-5
155.197
DSPYCWLYGXGJNJ-UHFFFAOYSA-N
11073648
CC(C)(C)OC(=O)NCC#C
C8H13NO2
MFCD07367245
n-boc-propargylamine, n-tert-butoxycarbonyl propargylamine, n-boc-prop-2-ynylamine, tert-butyl prop-2-yn-1-ylcarbamate, n-boc-propargyl amine, tert-butyl n-prop-2-yn-1-yl carbamate, tert-butyl prop-2-ynylcarbamate, n-boc-2-propyn-1-amine, n-boc propargyl amine, prop-2-ynyl-carbamic acid tert-butyl ester
tert-butyl N-prop-2-ynylcarbamate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

SDS Documents