Please login to your online account to display your discounted pricing

N-BOC-Piperidine-4-carbonitrile, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-BOC-Piperidine-4-carbonitrile
CAS 91419-52-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H18N2O2
Formula Weight 210.28
Synonym 1-BOC-4-cyanopiperidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC441740010

  SDS
thermo fisher scientific
AC441740010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC441740050

  SDS
thermo fisher scientific
AC441740050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.28
Physical Form Crystalline Powder
Assay ≥96.0%
Quantity 1g
Color White to off-white
Melting Point 57° to 62°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Synonym 1-BOC-4-cyanopiperidine
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 GHS Signal Word: Warning
CAS 91419-52-2
Molecular Formula C11H18N2O2
Chemical Name or Material N-BOC-Piperidine-4-carbonitrile
Assay Percent Range 97%
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.