missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-BOC-Piperidine-2-carboxylic acid, +97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC384760050

 View more versions of this product

Catalog No. AC384760050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.0%
Crystalline Powder
White
97.0
97+%
Glass bottle
N-BOC-2-Piperidinecarboxylic acid
229.28
5g
100.0
98303-20-9
MFCD01862877
Authentic
C11H19NO4

Chemical Identifiers

98303-20-9
100.0
C11H19NO4
MFCD01862877
≥97.0%
97.0
97+%
229.28
N-BOC-2-Piperidinecarboxylic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.