missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-O-methyl-L-threonine, 95%

$45.95 - $539.74

Chemical Identifiers

CAS 48068-25-3
Molecular Formula C10H19NO5
Molecular Weight (g/mol) 233.264
MDL Number MFCD00076990
InChI Key VWSUOKFUIPMDDX-RQJHMYQMSA-N
Synonym boc-o-methyl-l-threonine, boc-thr me-oh, 2s,3r-2-tert-butoxycarbonylamino-3-methoxybutanoic acid, 2s,3r-2-tert-butoxycarbonyl amino-3-methoxybutanoic acid, n-boc-o-methyl-l-threonine, o-methyl-l-threonine, n-boc protected, n-tert-butoxycarbonyl-o-methyl-l-threonine, n-tert-butyloxycarbonyl-o-methyl-l-threonine, 2s,3r-2-tert-butoxycarbonyl amino-3-methoxybutanoic acid, 2s,3r-2-tert-butoxy carbonyl amino-3-methoxybutanoic acid
PubChem CID 15460346
IUPAC Name (2S,3R)-3-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C(C(=O)O)NC(=O)OC(C)(C)C)OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63750MD View Documents Alfa Aesar™
H63750MD
250mg Each for $45.95
 
AAH6375003 View Documents Alfa Aesar™
H6375003
1g Each for $135.85
 
AAH6375006 View Documents Alfa Aesar™
H6375006
5g Each for $539.74
 
Specifications

Chemical Identifiers

48068-25-3
233.264
VWSUOKFUIPMDDX-RQJHMYQMSA-N
15460346
CC(C(C(=O)O)NC(=O)OC(C)(C)C)OC
C10H19NO5
MFCD00076990
boc-o-methyl-l-threonine, boc-thr me-oh, 2s,3r-2-tert-butoxycarbonylamino-3-methoxybutanoic acid, 2s,3r-2-tert-butoxycarbonyl amino-3-methoxybutanoic acid, n-boc-o-methyl-l-threonine, o-methyl-l-threonine, n-boc protected, n-tert-butoxycarbonyl-o-methyl-l-threonine, n-tert-butyloxycarbonyl-o-methyl-l-threonine, 2s,3r-2-tert-butoxycarbonyl amino-3-methoxybutanoic acid, 2s,3r-2-tert-butoxy carbonyl amino-3-methoxybutanoic acid
(2S,3R)-3-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid

Specifications

48068-25-3
MFCD00076990
VWSUOKFUIPMDDX-RQJHMYQMSA-N
(2S,3R)-3-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
15460346
Solid
250mg
C10H19NO5
boc-o-methyl-l-threonine, boc-thr me-oh, 2s,3r-2-tert-butoxycarbonylamino-3-methoxybutanoic acid, 2s,3r-2-tert-butoxycarbonyl amino-3-methoxybutanoic acid, n-boc-o-methyl-l-threonine, o-methyl-l-threonine, n-boc protected, n-tert-butoxycarbonyl-o-methyl-l-threonine, n-tert-butyloxycarbonyl-o-methyl-l-threonine, 2s,3r-2-tert-butoxycarbonyl amino-3-methoxybutanoic acid, 2s,3r-2-tert-butoxy carbonyl amino-3-methoxybutanoic acid
CC(C(C(=O)O)NC(=O)OC(C)(C)C)OC
233.264
233.26
95%
N-Boc-O-methyl-L-threonine
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C