missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-O-methyl-L-threonine, 95%

Manufacturer:  Alfa Aesar™ H63750MD

 View more versions of this product

Catalog No. AAH63750MD



Specifications

Specifications

N-Boc-O-methyl-L-threonine
C10H19NO5
boc-o-methyl-l-threonine, boc-thr me-oh, 2s,3r-2-tert-butoxycarbonylamino-3-methoxybutanoic acid, 2s,3r-2-tert-butoxycarbonyl amino-3-methoxybutanoic acid, n-boc-o-methyl-l-threonine, o-methyl-l-threonine, n-boc protected, n-tert-butoxycarbonyl-o-methyl-l-threonine, n-tert-butyloxycarbonyl-o-methyl-l-threonine, 2s,3r-2-tert-butoxycarbonyl amino-3-methoxybutanoic acid, 2s,3r-2-tert-butoxy carbonyl amino-3-methoxybutanoic acid
CC(C(C(=O)O)NC(=O)OC(C)(C)C)OC
233.264
233.26
95%
48068-25-3
MFCD00076990
VWSUOKFUIPMDDX-RQJHMYQMSA-N
(2S,3R)-3-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
15460346
Solid
250mg

Chemical Identifiers

48068-25-3
233.264
VWSUOKFUIPMDDX-RQJHMYQMSA-N
15460346
CC(C(C(=O)O)NC(=O)OC(C)(C)C)OC
C10H19NO5
MFCD00076990
boc-o-methyl-l-threonine, boc-thr me-oh, 2s,3r-2-tert-butoxycarbonylamino-3-methoxybutanoic acid, 2s,3r-2-tert-butoxycarbonyl amino-3-methoxybutanoic acid, n-boc-o-methyl-l-threonine, o-methyl-l-threonine, n-boc protected, n-tert-butoxycarbonyl-o-methyl-l-threonine, n-tert-butyloxycarbonyl-o-methyl-l-threonine, 2s,3r-2-tert-butoxycarbonyl amino-3-methoxybutanoic acid, 2s,3r-2-tert-butoxy carbonyl amino-3-methoxybutanoic acid
(2S,3R)-3-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents