Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-O-ethyl-D-tyrosine, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-O-ethyl-D-tyrosine
CAS 76757-92-1
Formula Weight 309.36
MDL Number MFCD00065602
Beilstein 3622828
Synonym Boc-D-Tyr(Et)-OH; (R)-Boc-2-amino-3-(4-ethoxyphenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6617103

 
alfa aesar™
H6617103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66171MD

 
alfa aesar™
H66171MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 309.36
Chemical Name or Material N-Boc-O-ethyl-D-tyrosine
CAS 76757-92-1
Assay 95%
MDL Number MFCD00065602
Beilstein 3622828
Synonym Boc-D-Tyr(Et)-OH; (R)-Boc-2-amino-3-(4-ethoxyphenyl)propionic acid
Quantity 1g
Storage Note 1 -30° to -10°C
Physical Form Solid
Safety and Handling

-30° to -10°C