missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-O-benzyl-L-tyrosine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2345514
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Boc-O-benzyl-L-tyrosine | |
108°C to 110°C | |
MFCD00065597 | |
boc-tyr bzl-oh, boc-o-benzyl-l-tyrosine, n-boc-o-benzyl-l-tyrosine, n-tert-butoxycarbonyl-o-benzyl-l-tyrosine, s-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-butoxycarbonyl-o-benzyl-l-tyrisine, o-benzyl-n-tert-butoxycarbonyl-l-tyrosine, 2s-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, o-benzyl-n-tert-butoxy carbonyl-l-tyrosine, tyrosine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl | |
CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O | |
371.43 | |
371.43 |
2130-96-3 | |
C21H25NO5 | |
25 g | |
ZAVSPTOJKOFMTA-LGWFVXIRNA-N | |
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(4-phenylmethoxyphenyl)propanoic acid | |
98763 | |
98% |
Chemical Identifiers
2130-96-3 | |
371.43 | |
ZAVSPTOJKOFMTA-LGWFVXIRNA-N | |
98763 | |
CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O |
C21H25NO5 | |
MFCD00065597 | |
boc-tyr bzl-oh, boc-o-benzyl-l-tyrosine, n-boc-o-benzyl-l-tyrosine, n-tert-butoxycarbonyl-o-benzyl-l-tyrosine, s-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-butoxycarbonyl-o-benzyl-l-tyrisine, o-benzyl-n-tert-butoxycarbonyl-l-tyrosine, 2s-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, o-benzyl-n-tert-butoxy carbonyl-l-tyrosine, tyrosine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl | |
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(4-phenylmethoxyphenyl)propanoic acid |
Safety and Handling
EINECSNumber : 218-349-4
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only