missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-BOC-O-Benzyl-L-serine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC334660250

 View more versions of this product

Catalog No. AC334660250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
Powder
White
21.50 (20.00°C c=2,ethanol)
+19° to +24° (20°C, 589nm) (c=2, EtOH)
C15H21NO5
N-BOC-O-Benzyl-L-serine
98%
Authentic
295.33
25g
58° to 63°C
100.0
MFCD00066063
23680-31-1
97.5
98%
Glass bottle

Chemical Identifiers

23680-31-1
100.0
C15H21NO5
MFCD00066063
≥97.5%
97.5
98%
295.33
N-BOC-O-Benzyl-L-serine, 98%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 245-820-1

TSCA : TSCA