Please login to your online account to display your discounted pricing

N-BOC-O-Benzyl-L-serine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-BOC-O-Benzyl-L-serine
Name Note 98%
CAS 23680-31-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C15H21NO5
Formula Weight 295.33
MDL Number MFCD00066063
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC334660250

  SDS
thermo fisher scientific
AC334660250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 295.33
Quantity 25g
Color White
EINECS Number 245-820-1
TSCA TSCA
Specific Rotation Condition 21.50 (20.00°C c=2,ethanol)
Melting Point 58° to 63°C
CAS Max % 100.0
Specific Rotation +19° to +24° (20°C, 589nm) (c=2, EtOH)
MDL Number MFCD00066063
Molecular Formula C15H21NO5
Chemical Name or Material N-BOC-O-Benzyl-L-serine
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
CAS 23680-31-1
CAS Min % 97.5
Infrared Spectrum Authentic