missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-O-benzyl-L-serine, 97%

Manufacturer:  Alfa Aesar™ H6270522

 View more versions of this product

Catalog No. AAH6270522



Specifications

Specifications

N-Boc-O-benzyl-L-serine
C15H21NO5
3064461
DMBKPDOAQVGTST-LBPRGKRZSA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxypropanoic acid
90234
Solid
58°C to 60°C
23680-31-1
MFCD00066063
2s-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylmethoxypropanoic acid, 2s-3-benzyl-oxy-2-tert-butoxy carbonyl amino propanoic acid, s-3-benzyloxy-2-tert-butoxycarbonyl amino propanoic acid, 2s-3-benzyloxy-2-tert-butoxycarbonyl amino propanoic acid, s-3-benzyloxy-2-tert-butoxycarbonylamino propanoic acid, l-serine n-11-dimethylethoxy carbonyl-o-phenylmethyl, 2s-3-benzyloxy-2-tert-butoxycarbonylamino propionic acid, n-11-dimethylethoxy carbonyl-o-phenylmethyl-l-serine
CC(C)(C)OC(=O)NC(COCC1=CC=CC=C1)C(=O)O
295.335
295.33
97%
100g

Chemical Identifiers

23680-31-1
295.335
DMBKPDOAQVGTST-LBPRGKRZSA-N
90234
CC(C)(C)OC(=O)NC(COCC1=CC=CC=C1)C(=O)O
C15H21NO5
MFCD00066063
2s-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylmethoxypropanoic acid, 2s-3-benzyl-oxy-2-tert-butoxy carbonyl amino propanoic acid, s-3-benzyloxy-2-tert-butoxycarbonyl amino propanoic acid, 2s-3-benzyloxy-2-tert-butoxycarbonyl amino propanoic acid, s-3-benzyloxy-2-tert-butoxycarbonylamino propanoic acid, l-serine n-11-dimethylethoxy carbonyl-o-phenylmethyl, 2s-3-benzyloxy-2-tert-butoxycarbonylamino propionic acid, n-11-dimethylethoxy carbonyl-o-phenylmethyl-l-serine
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxypropanoic acid
Documents