missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-O-benzyl-D-threonine, 98%

$72.97 - $250.73

Chemical Identifiers

CAS 69355-99-3
Molecular Formula C16H23NO5
Molecular Weight (g/mol) 309.362
MDL Number MFCD00038247
InChI Key CTXPLTPDOISPTE-WCQYABFASA-N
Synonym 2r,3s-3-benzyloxy-2-tert-butoxycarbonyl amino butanoic acid
PubChem CID 6992573
IUPAC Name (2R,3S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxybutanoic acid
SMILES CC(C(C(=O)O)NC(=O)OC(C)(C)C)OCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6148503 View Documents Alfa Aesar™
H6148503
1g Each for $72.97
 
AAH6148506 View Documents Alfa Aesar™
H6148506
5g Each for $250.73
 
Specifications

Chemical Identifiers

69355-99-3
309.362
CTXPLTPDOISPTE-WCQYABFASA-N
6992573
CC(C(C(=O)O)NC(=O)OC(C)(C)C)OCC1=CC=CC=C1
C16H23NO5
MFCD00038247
2r,3s-3-benzyloxy-2-tert-butoxycarbonyl amino butanoic acid
(2R,3S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxybutanoic acid

Specifications

69355-99-3
MFCD00038247
2r,3s-3-benzyloxy-2-tert-butoxycarbonyl amino butanoic acid
CC(C(C(=O)O)NC(=O)OC(C)(C)C)OCC1=CC=CC=C1
309.362
309.36
98%
1g
C16H23NO5
7339503
CTXPLTPDOISPTE-WCQYABFASA-N
(2R,3S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxybutanoic acid
6992573
Solid
115°C to 118°C
N-Boc-O-benzyl-D-threonine
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures