missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-O-benzyl-D-threonine, 98%

Manufacturer:  Alfa Aesar™ H6148503

 View more versions of this product

Catalog No. AAH6148503



Specifications

Specifications

N-Boc-O-benzyl-D-threonine
C16H23NO5
7339503
CTXPLTPDOISPTE-WCQYABFASA-N
(2R,3S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxybutanoic acid
6992573
Solid
115°C to 118°C
69355-99-3
MFCD00038247
2r,3s-3-benzyloxy-2-tert-butoxycarbonyl amino butanoic acid
CC(C(C(=O)O)NC(=O)OC(C)(C)C)OCC1=CC=CC=C1
309.362
309.36
98%
1g

Chemical Identifiers

69355-99-3
309.362
CTXPLTPDOISPTE-WCQYABFASA-N
6992573
CC(C(C(=O)O)NC(=O)OC(C)(C)C)OCC1=CC=CC=C1
C16H23NO5
MFCD00038247
2r,3s-3-benzyloxy-2-tert-butoxycarbonyl amino butanoic acid
(2R,3S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxybutanoic acid
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents