Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-O-benzyl-D-serine, 99%

$85.14 - $85.14

Chemical Identifiers

Chemical Name or Material N-Boc-O-benzyl-D-serine
CAS 47173-80-8
Molecular Formula C15H21NO5
Formula Weight 295.31
MDL Number MFCD00038248
Synonym Boc-D-Ser(Bzl)-OH; N-tert-Butoxycarbonyl-O-benzyl-D-serine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2266603

  SDS
alfa aesar™
B2266603
1g Each for $85.14
Description & Specifications

Specifications

Formula Weight 295.31
MDL Number MFCD00038248
Synonym Boc-D-Ser(Bzl)-OH; N-tert-Butoxycarbonyl-O-benzyl-D-serine
Chemical Name or Material N-Boc-O-benzyl-D-serine
Quantity 1g
Melting Point 60° to 63°C
Assay 99%
Molecular Formula C15H21NO5
CAS 47173-80-8