missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-BOC-O-Benzyl-D-serine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 334670010

 View more versions of this product

Catalog No. AC334670010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

N-BOC-O-Benzyl-D-serine
47173-80-8
100.0
Glass bottle
-20° to -24° (20°C, 589nm) (c=1, EtOH)
98%
58° to 63°C
MFCD00038248
White
98%
97.5
Authentic
Powder
≥97.5%
C15H21NO5
295.33
1g

Chemical Identifiers

47173-80-8
100.0
C15H21NO5
MFCD00038248
≥97.5%
97.5
98%
295.33
N-BOC-O-Benzyl-D-serine, 98%