Please login to your online account to display your discounted pricing

Nα-BOC-Nepsilon-CBZ-L-Lysine DCHA, 99.5+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  275620050

 View more versions of this product

Catalog No. AC275620050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.50%
Physical Form Fine Crystalline Powder
Formula Weight 561.76
Quantity 5g
Color White
EINECS Number 241-018-0
Melting Point 113° to 116°C
CAS Max % 100.0
Packaging Glass bottle
Name Note 99.5+%
MDL Number MFCD00066511
Infrared Spectrum Authentic
Chemical Name or Material Nα-BOC-Nε-CBZ-L-Lysine DCHA
Assay Percent Range 99.5+%
Molecular Formula C19H28N2O6·C12H23N
CAS Min % 99.5
Specific Rotation +7.5° to +9.5° (20°C, 589nm) (c=2, CH3OH)
CAS 16948-04-2

Specifications

Chemical Name or Material Nα-BOC-Nε-CBZ-L-Lysine DCHA
Name Note 99.5+%
CAS 16948-04-2
CAS Min % 99.5
CAS Max % 100.0
Assay ≥99.50%
Assay Percent Range 99.5+%
Molecular Formula C19H28N2O6·C12H23N
Formula Weight 561.76
MDL Number MFCD00066511