Please login to your online account to display your discounted pricing

Nα-BOC-Nepsilon-CBZ-L-Lysine DCHA, 99.5+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Nα-BOC-Nε-CBZ-L-Lysine DCHA
Name Note 99.5+%
CAS 16948-04-2
CAS Min % 99.5
CAS Max % 100.0
Assay ≥99.50%
Assay Percent Range 99.5+%
Molecular Formula C19H28N2O6·C12H23N
Formula Weight 561.76
MDL Number MFCD00066511
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC275620050

  SDS
thermo fisher scientific
275620050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.50%
Physical Form Fine Crystalline Powder
Formula Weight 561.76
Quantity 5g
Color White
EINECS Number 241-018-0
Melting Point 113° to 116°C
CAS Max % 100.0
Packaging Glass bottle
Name Note 99.5+%
MDL Number MFCD00066511
Infrared Spectrum Authentic
Chemical Name or Material Nα-BOC-Nε-CBZ-L-Lysine DCHA
Assay Percent Range 99.5+%
Molecular Formula C19H28N2O6·C12H23N
CAS Min % 99.5
Specific Rotation +7.5° to +9.5° (20°C, 589nm) (c=2, CH3OH)
CAS 16948-04-2