missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-α-BOC-N-β-Xanthenyl-L-asparagine, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC440840050

 View more versions of this product

Catalog No. AC440840050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(0.5mmol/mL DMF) clear, colorless
Authentic
1% Max.
170°C to 172°C
65420-40-8
100.0
C22H24N2O6
YMGDQLXBNMRJMR-HNNXBMFYSA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-(9H-xanthen-9-ylamino)butanoic acid
7017974
Powder
N-α-BOC-N-β-Xanthenyl-L-asparagine
9.5
White
5g
95.0
95% min. (HPLC)
2s-2-tert-butoxycarbonyl amino-3-9h-xanthen-9-yl carbamoyl propanoic acid, ambotzbaa1070, boc-asn xan-oh, n alpha-boc-n gamma-9-xanthenyl-l-asparagine, n-alpha-t-boc-n-beta-xanthyl-l-asparagine, n-boc-n-xanthyl-l-asparagine, nalpha-boc-nbeta-xanthenyl-l-asparagine, pubchem18926, s-4-9h-xanthen-9-yl amino-2-tert-butoxycarbonyl amino-4-oxobutanoic acid
CC(C)(C)OC(=O)NC(CC(=O)NC1C2=CC=CC=C2OC3=CC=CC=C13)C(=O)O
412.45
412.45
96%

Chemical Identifiers

65420-40-8
412.45
2s-2-tert-butoxycarbonyl amino-3-9h-xanthen-9-yl carbamoyl propanoic acid, ambotzbaa1070, boc-asn xan-oh, n alpha-boc-n gamma-9-xanthenyl-l-asparagine, n-alpha-t-boc-n-beta-xanthyl-l-asparagine, n-boc-n-xanthyl-l-asparagine, nalpha-boc-nbeta-xanthenyl-l-asparagine, pubchem18926, s-4-9h-xanthen-9-yl amino-2-tert-butoxycarbonyl amino-4-oxobutanoic acid
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-(9H-xanthen-9-ylamino)butanoic acid
C22H24N2O6
YMGDQLXBNMRJMR-HNNXBMFYSA-N
7017974
CC(C)(C)OC(=O)NC(CC(=O)NC1C2=CC=CC=C2OC3=CC=CC=C13)C(=O)O
SDS Documents