missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-α-BOC-N-omega-Tosyl-L-arginine, 97%, contains <15% ethyl acetate, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC440830250

 View more versions of this product

Catalog No. AC440830250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-α-BOC-N-ω-Tosyl-L-arginine
15.0% max. (EtOAc)
1% Max.
99°C to 100°C
13836-37-8,141-78-6
100.0
C18H28N4O6S
WBIIPXYJAMICNU-AWEZNQCLSA-N
(2S)-5-[[amino-[(4-methylphenyl)sulfonylamino]methylidene]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
83767
Powder
Authentic
-3.2
White
25g
96.0
96% min. (HPLC)
2s-2-tert-butoxycarbonyl amino-5-n'-4-methylbenzenesulfonyl carbamimidamido pentanoic acid, ambotzbaa1068, boc arg tos-oh, boc-arg tos, boc-arg tos oh, boc-arg tos-oh, boc-arg ts, l-ornithine, n2-1,1-dimethylethoxy carbonyl-n5-imino 4-methylphenyl sulfonyl amino methyl, n 2-tert-butoxycarbonyl-n g-tosyl-l-arginine, n alpha-boc-n omega-tosyl-l-arginine
CC1=CC=C(C=C1)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OC(C)(C)C)N
428.51
428.51
97%

Chemical Identifiers

13836-37-8
428.51
2s-2-tert-butoxycarbonyl amino-5-n'-4-methylbenzenesulfonyl carbamimidamido pentanoic acid, ambotzbaa1068, boc arg tos-oh, boc-arg tos, boc-arg tos oh, boc-arg tos-oh, boc-arg ts, l-ornithine, n2-1,1-dimethylethoxy carbonyl-n5-imino 4-methylphenyl sulfonyl amino methyl, n 2-tert-butoxycarbonyl-n g-tosyl-l-arginine, n alpha-boc-n omega-tosyl-l-arginine
(2S)-5-[[amino-[(4-methylphenyl)sulfonylamino]methylidene]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
C18H28N4O6S
WBIIPXYJAMICNU-AWEZNQCLSA-N
83767
CC1=CC=C(C=C1)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OC(C)(C)C)N
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/equipment.
Wear protective gloves/protective cl

GHS Signal Word: Warning

EINECSNumber : 237-549-2

SDS Documents