Please login to your online account to display your discounted pricing

N-BOC-N-Methylethylenediamine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-BOC-N-Methylethylenediamine
CAS 121492-06-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H18N2O2
Formula Weight 174.24
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC437340050

  SDS
thermo fisher scientific
AC437340050
Glass bottle 5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC437340010

  SDS
thermo fisher scientific
AC437340010
Glass bottle 1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 174.24
Physical Form Liquid
Quantity 5mL
Color Clear colorless to pale yellow
Density 0.975g/mL
CAS Max % 100.0
CAS Min % 96.0
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
CAS 121492-06-6
Assay Percent Range 97%
Molecular Formula C8H18N2O2
Chemical Name or Material N-BOC-N-Methylethylenediamine
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.