missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-N-methyl-O-benzyl-D-tyrosine, 95%

$119.05 - $380.91

Chemical Identifiers

CAS 138774-98-8
Molecular Formula C22H27NO5
Molecular Weight (g/mol) 385.46
MDL Number MFCD02259475
InChI Key FSNRGORPOYPIJC-LJQANCHMSA-N
Synonym boc-d-metyr bzl-oh, boc-n-me-d-tyr bzl-oh, d-tyrosine, n-1, 1-dimethylethoxy carbonyl-n-methyl-o-phenylmethyl, 2r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl methyl amino propanoic acid, ambotzbaa5560, boc-d-n-me-tyr bzl-oh, boc-n-methyl-o-benzyl-d-tyrosine, boc-nalpha-methyl-o-benzyl-d-tyrosine, n-alpha-t-butyloxycarbonyl-n-alpha-methyl-o-benzyl-d-tyrosine, d-tyrosine,n-1,1-dimethylethoxy carbonyl-n-methyl-o-phenylmethyl
PubChem CID 7010683
IUPAC Name (2R)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-3-(4-phenylmethoxyphenyl)propanoic acid
SMILES CC(C)(C)OC(=O)N(C)C(CC1=CC=C(C=C1)OCC2=CC=CC=C2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6620003 View Documents Alfa Aesar™
H6620003
1g Each for $380.91
 
AAH66200MD View Documents Alfa Aesar™
H66200MD
250mg Each for $119.05
 
Specifications

Chemical Identifiers

138774-98-8
385.46
FSNRGORPOYPIJC-LJQANCHMSA-N
7010683
CC(C)(C)OC(=O)N(C)C(CC1=CC=C(C=C1)OCC2=CC=CC=C2)C(=O)O
C22H27NO5
MFCD02259475
boc-d-metyr bzl-oh, boc-n-me-d-tyr bzl-oh, d-tyrosine, n-1, 1-dimethylethoxy carbonyl-n-methyl-o-phenylmethyl, 2r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl methyl amino propanoic acid, ambotzbaa5560, boc-d-n-me-tyr bzl-oh, boc-n-methyl-o-benzyl-d-tyrosine, boc-nalpha-methyl-o-benzyl-d-tyrosine, n-alpha-t-butyloxycarbonyl-n-alpha-methyl-o-benzyl-d-tyrosine, d-tyrosine,n-1,1-dimethylethoxy carbonyl-n-methyl-o-phenylmethyl
(2R)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-3-(4-phenylmethoxyphenyl)propanoic acid

Specifications

138774-98-8
MFCD02259475
FSNRGORPOYPIJC-LJQANCHMSA-N
(2R)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-3-(4-phenylmethoxyphenyl)propanoic acid
7010683
Solid
1g
C22H27NO5
boc-d-metyr bzl-oh, boc-n-me-d-tyr bzl-oh, d-tyrosine, n-1, 1-dimethylethoxy carbonyl-n-methyl-o-phenylmethyl, 2r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl methyl amino propanoic acid, ambotzbaa5560, boc-d-n-me-tyr bzl-oh, boc-n-methyl-o-benzyl-d-tyrosine, boc-nalpha-methyl-o-benzyl-d-tyrosine, n-alpha-t-butyloxycarbonyl-n-alpha-methyl-o-benzyl-d-tyrosine, d-tyrosine,n-1,1-dimethylethoxy carbonyl-n-methyl-o-phenylmethyl
CC(C)(C)OC(=O)N(C)C(CC1=CC=C(C=C1)OCC2=CC=CC=C2)C(=O)O
385.46
385.46
95%
N-Boc-N-methyl-O-benzyl-D-tyrosine
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C