missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-N-methyl-L-serine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6331406
Description
N-Boc-N-methyl-L-serine is used as a precursor in organic synthesis and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-N-methyl-L-serine is used as a precursor in organic synthesis and pharmaceuticals.
Notes
Incompatible with strong oxidizing agents.
Specifications
83°C to 87°C | |
101772-29-6 | |
MFCD00037249 | |
boc-n-me-ser-oh, boc-l-meser-oh, boc-n-a-methyl-l-serine, boc-n-a-methyl-l-serine, boc-n-, s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, l-serine,n-1,1-dimethylethoxy carbonyl-n-methyl, boc-meser-oh, 2s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, 2s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid | |
CC(C)(C)OC(=O)N(C)C(CO)C(=O)O | |
219.237 | |
219.23 | |
5 g |
N-Boc-N-methyl-L-serine | |
C9H17NO5 | |
8685334 | |
NJQGIDVCNBZXLG-LURJTMIESA-N | |
(2S)-3-hydroxy-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid | |
7009594 | |
98% | |
Solid |
Chemical Identifiers
101772-29-6 | |
219.237 | |
NJQGIDVCNBZXLG-LURJTMIESA-N | |
7009594 | |
CC(C)(C)OC(=O)N(C)C(CO)C(=O)O |
C9H17NO5 | |
MFCD00037249 | |
boc-n-me-ser-oh, boc-l-meser-oh, boc-n-a-methyl-l-serine, boc-n-a-methyl-l-serine, boc-n-, s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, l-serine,n-1,1-dimethylethoxy carbonyl-n-methyl, boc-meser-oh, 2s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, 2s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid | |
(2S)-3-hydroxy-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid |
Safety and Handling
TSCA : No
RUO – Research Use Only