missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-N-methyl-L-phenylglycine, 97%, Thermo Scientific™
$45.84 - $135.21
Chemical Identifiers
CAS | 30925-11-2 |
---|---|
Molecular Formula | C14H19NO4 |
Molecular Weight (g/mol) | 265.31 |
MDL Number | MFCD00038799 |
InChI Key | COABPHLHHQAKPL-UHFFFAOYNA-N |
Synonym | boc-n-me-phg-oh, boc-n-methyl-l-phenylglycine, s-tert-butoxycarbonyl methyl amino phenyl acetic acid, s-2-tert-butoxycarbonyl methyl amino-2-phenylacetic acid, ambotzbaa1259, pubchem12248, boc-n-methyl-s-2-phenylglycine, s-methyl tert-butoxycarbonyl amino phenylacetic acid, n-alpha-t-butyloxycarbonyl-n-alpha-methyl-l-phenylglycine, 2s-tert-butoxycarbonyl methyl amino phenyl acetic acid |
PubChem CID | 7010620 |
SMILES | CN(C(C(O)=O)C1=CC=CC=C1)C(=O)OC(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH63720MD
|
Thermo Scientific Chemicals
H63720MD |
250 mg |
Each for $45.84
|
|
|||||
AAH6372003
|
Thermo Scientific Chemicals
H6372003 |
1 g |
Each for $135.21
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
30925-11-2 | |
265.31 | |
COABPHLHHQAKPL-UHFFFAOYNA-N | |
7010620 |
C14H19NO4 | |
MFCD00038799 | |
boc-n-me-phg-oh, boc-n-methyl-l-phenylglycine, s-tert-butoxycarbonyl methyl amino phenyl acetic acid, s-2-tert-butoxycarbonyl methyl amino-2-phenylacetic acid, ambotzbaa1259, pubchem12248, boc-n-methyl-s-2-phenylglycine, s-methyl tert-butoxycarbonyl amino phenylacetic acid, n-alpha-t-butyloxycarbonyl-n-alpha-methyl-l-phenylglycine, 2s-tert-butoxycarbonyl methyl amino phenyl acetic acid | |
CN(C(C(O)=O)C1=CC=CC=C1)C(=O)OC(C)(C)C |
Specifications
30925-11-2 | |
MFCD00038799 | |
COABPHLHHQAKPL-UHFFFAOYNA-N | |
(2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-2-phenylacetic acid | |
7010620 | |
97% | |
Solid |
C14H19NO4 | |
boc-n-me-phg-oh, boc-n-methyl-l-phenylglycine, s-tert-butoxycarbonyl methyl amino phenyl acetic acid, s-2-tert-butoxycarbonyl methyl amino-2-phenylacetic acid, ambotzbaa1259, pubchem12248, boc-n-methyl-s-2-phenylglycine, s-methyl tert-butoxycarbonyl amino phenylacetic acid, n-alpha-t-butyloxycarbonyl-n-alpha-methyl-l-phenylglycine, 2s-tert-butoxycarbonyl methyl amino phenyl acetic acid | |
CN(C(C(O)=O)C1=CC=CC=C1)C(=O)OC(C)(C)C | |
265.31 | |
265.3 | |
250 mg | |
N-Boc-N-methyl-L-phenylglycine |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold