missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-N-methyl-L-phenylglycine, 97%

Manufacturer:  Alfa Aesar™ H6372006

 View more versions of this product

Catalog No. AAH6372006



Specifications

Specifications

N-Boc-N-methyl-L-phenylglycine
C14H19NO4
boc-n-me-phg-oh, boc-n-methyl-l-phenylglycine, s-tert-butoxycarbonyl methyl amino phenyl acetic acid, s-2-tert-butoxycarbonyl methyl amino-2-phenylacetic acid, ambotzbaa1259, pubchem12248, boc-n-methyl-s-2-phenylglycine, s-methyl tert-butoxycarbonyl amino phenylacetic acid, n-alpha-t-butyloxycarbonyl-n-alpha-methyl-l-phenylglycine, 2s-tert-butoxycarbonyl methyl amino phenyl acetic acid
CC(C)(C)OC(=O)N(C)C(C1=CC=CC=C1)C(=O)O
265.309
265.3
97%
30925-11-2
MFCD00038799
COABPHLHHQAKPL-NSHDSACASA-N
(2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-2-phenylacetic acid
7010620
Solid
5g

Chemical Identifiers

30925-11-2
265.309
COABPHLHHQAKPL-NSHDSACASA-N
7010620
CC(C)(C)OC(=O)N(C)C(C1=CC=CC=C1)C(=O)O
C14H19NO4
MFCD00038799
boc-n-me-phg-oh, boc-n-methyl-l-phenylglycine, s-tert-butoxycarbonyl methyl amino phenyl acetic acid, s-2-tert-butoxycarbonyl methyl amino-2-phenylacetic acid, ambotzbaa1259, pubchem12248, boc-n-methyl-s-2-phenylglycine, s-methyl tert-butoxycarbonyl amino phenylacetic acid, n-alpha-t-butyloxycarbonyl-n-alpha-methyl-l-phenylglycine, 2s-tert-butoxycarbonyl methyl amino phenyl acetic acid
(2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-2-phenylacetic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents