Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-N-methyl-D-valine, 95%

$30.16 - $30.16

Chemical Identifiers

Chemical Name or Material N-Boc-N-methyl-D-valine
CAS 89536-85-6
Formula Weight 231.29
MDL Number MFCD00076996
Synonym Boc-N-Me-D-Val-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6244303

 
alfa aesar™
H6244303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6244306

 
alfa aesar™
H6244306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62443MD

 
alfa aesar™
H62443MD
250mg Each for $30.16
Description & Specifications

Specifications

Formula Weight 231.29
Chemical Name or Material N-Boc-N-methyl-D-valine
Physical Form Solid
Assay 95%
MDL Number MFCD00076996
CAS 89536-85-6
Synonym Boc-N-Me-D-Val-OH
Quantity 1g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C