missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-N-methyl-D-valine, 95%

Manufacturer:  Alfa Aesar™ H6244306

 View more versions of this product

Catalog No. AAH6244306



Specifications

Specifications

N-Boc-N-methyl-D-valine
C11H21NO4
2r-2-tert-butoxycarbonyl methyl amino-3-methylbutanoic acid
CC(C)C(C(=O)O)N(C)C(=O)OC(C)(C)C
231.292
231.29
95%
89536-85-6
MFCD00076996
XPUAXAVJMJDPDH-MRVPVSSYSA-N
(2R)-3-methyl-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]butanoic acid
7018800
Solid
5g

Chemical Identifiers

89536-85-6
231.292
XPUAXAVJMJDPDH-MRVPVSSYSA-N
7018800
CC(C)C(C(=O)O)N(C)C(=O)OC(C)(C)C
C11H21NO4
MFCD00076996
2r-2-tert-butoxycarbonyl methyl amino-3-methylbutanoic acid
(2R)-3-methyl-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]butanoic acid
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

Documents