missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-BOC-N,2-Dimethylalanine, 98+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC370950010
Specifications
N-BOC-N, 2-Dimethylalanine | |
Glass bottle | |
153°C to 157°C | |
146000-39-7 | |
100.0 | |
C10H19NO4 | |
MFCD01862912 | |
KPDLITGXUYMJEC-UHFFFAOYSA-N | |
2-methyl-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid | |
2734752 | |
Crystalline Powder |
Authentic | |
White | |
1g | |
98.0 | |
98+% | |
(CH3)3COCON(CH3)C(CH3)2COOH | |
2-methyl-2-methyl-2-methylpropan-2-yl oxycarbonyl amino propanoic acid, 2-tert-butoxycarbonyl methyl amino-2-methylpropanoic acid, a-methylamino isobutyric acid, acmc-1c0qg, boc-n,2-dimethylalanine, boc-n-me-aib-oh, boc-n-methyl-aib, boc-n-methyl-aminoisobutyric acid, n-boc-, n-boc-n,2-dimethyl-dl-alanine, n-boc-n,2-dimethylalanine | |
CC(C)(C)OC(=O)N(C)C(C)(C)C(=O)O | |
217.26 | |
217.26 | |
98+% |
Chemical Identifiers
146000-39-7 | |
217.26 | |
KPDLITGXUYMJEC-UHFFFAOYSA-N | |
2734752 | |
CC(C)(C)OC(=O)N(C)C(C)(C)C(=O)O |
C10H19NO4 | |
MFCD01862912 | |
2-methyl-2-methyl-2-methylpropan-2-yl oxycarbonyl amino propanoic acid, 2-tert-butoxycarbonyl methyl amino-2-methylpropanoic acid, a-methylamino isobutyric acid, acmc-1c0qg, boc-n,2-dimethylalanine, boc-n-me-aib-oh, boc-n-methyl-aib, boc-n-methyl-aminoisobutyric acid, n-boc-, n-boc-n,2-dimethyl-dl-alanine, n-boc-n,2-dimethylalanine | |
2-methyl-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid |
Safety and Handling
Recommended Storage : Refrigerator +4°C