Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-valinol, 97%

$29.69 - $79.41

Chemical Identifiers

Chemical Name or Material N-Boc-L-valinol
CAS 79069-14-0
Molecular Formula C10H21NO3
Formula Weight 203.28
MDL Number MFCD00082635
Beilstein 4663728
Synonym Boc-Val-ol; N-tert-Butoxycarbonyl-(S)-2-amino-3-methyl-1-butanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1023603

  SDS
alfa aesar™
L1023603
1g Each for $79.41

AAL10236MD

  SDS
alfa aesar™
L10236MD
250mg Each for $29.69
Description & Specifications

Specifications

Formula Weight 203.28
Chemical Name or Material N-Boc-L-valinol
MDL Number MFCD00082635
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
CAS 79069-14-0
Synonym Boc-Val-ol; N-tert-Butoxycarbonyl-(S)-2-amino-3-methyl-1-butanol
Beilstein 4663728
Optical Rotation +22° (c=1 in Chloroform)
Flash Point 85°C (185°F)
Health Hazard 1 Warning
Recommended Storage 2° to 8°C
Quantity 1g
Boiling Point 218°C
Molecular Formula C10H21NO3
Assay 97%
Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

2° to 8°C