Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-valine N-succinimidyl ester, 97%

Chemical Identifiers

Chemical Name or Material N-Boc-L-valine N-succinimidyl ester
CAS 3392-12-9
Formula Weight 314.34
MDL Number MFCD00037906
Beilstein 1551859
Synonym Boc-Val-OSu; N-Boc-L-valine N-hydroxysuccinimide ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6655914

  SDS
alfa aesar™
H6655914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6655922

  SDS
alfa aesar™
H6655922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 314.34
Assay 97%
Chemical Name or Material N-Boc-L-valine N-succinimidyl ester
MDL Number MFCD00037906
Synonym Boc-Val-OSu; N-Boc-L-valine N-hydroxysuccinimide ester
Color White
CAS 3392-12-9
Quantity 25g
Sensitivity Moisture Sensitive
Storage Note 1 -30° to -10°C
Physical Form Powder
Beilstein 1551859
Safety and Handling

-30° to -10°C