Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-valine methyl ester, 97%

Chemical Identifiers

Chemical Name or Material N-Boc-L-valine methyl ester
CAS 58561-04-9
Formula Weight 231.29
MDL Number MFCD00672510
Synonym Boc-Val-OMe
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6110103

  SDS
alfa aesar™
H6110103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6110106

  SDS
alfa aesar™
H6110106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 231.29
Chemical Name or Material N-Boc-L-valine methyl ester
Physical Form Liquid
Assay 97%
MDL Number MFCD00672510
CAS 58561-04-9
Synonym Boc-Val-OMe
Refractive Index 1.4400
Density 1.004g/mL
Melting Point 194°C
Quantity 1g