Please login to your online account to display your discounted pricing

N-α-BOC-L-tryptophanol, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC376700050

 View more versions of this product

Catalog No. AC376700050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 290.36
Physical Form Powder
Assay ≥97.5%
Quantity 5g
Color White to off-white
Melting Point 118° to 122°C
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material N-α-BOC-L-tryptophanol
Assay Percent Range 98%
Molecular Formula C16H22N2O3
MDL Number MFCD00235953
CAS 82689-19-8
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Specific Rotation -28° to -32° (20°C, 589nm) (C=1, Methanol)
Synonym N-alpha-(tert-Butoxycarbonyl)-L-tryptophanol
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material N-α-BOC-L-tryptophanol
CAS 82689-19-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C16H22N2O3
Formula Weight 290.36
MDL Number MFCD00235953
Synonym N-alpha-(tert-Butoxycarbonyl)-L-tryptophanol
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents