Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-tryptophan benzyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-L-tryptophan benzyl ester
CAS 57229-67-1
Formula Weight 394.47
MDL Number MFCD00190837
Synonym Boc-Trp-Obzl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6661103

 
alfa aesar™
H6661103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6661106

 
alfa aesar™
H6661106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6661114

 
alfa aesar™
H6661114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 140° to 146°C
Formula Weight 394.47
Chemical Name or Material N-Boc-L-tryptophan benzyl ester
MDL Number MFCD00190837
CAS 57229-67-1
Synonym Boc-Trp-Obzl
Quantity 1g
Assay 95%
Physical Form Powder
Storage Note 1 -30° to -10°C
Color Off-white to white
Safety and Handling

-30° to -10°C