Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-threonine, 98+%

$18.47 - $60.80

Chemical Identifiers

Chemical Name or Material N-Boc-L-threonine
CAS 2592-18-9
Molecular Formula C9H17NO5
Formula Weight 219.24
MDL Number MFCD00065946
Beilstein 2331474
Synonym Boc-Thr-OH; N-(tert-Butoxycarbonyl)-L-threonine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0911103

 
alfa aesar™
L0911103
1g Each for $18.47

AAL0911106

 
alfa aesar™
L0911106
5g Each for $60.80
Description & Specifications

Specifications

Formula Weight 219.24
Chemical Name or Material N-Boc-L-threonine
Assay ≥98%
Molecular Formula C9H17NO5
MDL Number MFCD00065946
Synonym Boc-Thr-OH; N-(tert-Butoxycarbonyl)-L-threonine
Melting Point 77° to 81°C
CAS 2592-18-9
Beilstein 2331474
Quantity 1g
Optical Rotation −8.5° (c=1 in Acetic Acid)
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C