Please login to your online account to display your discounted pricing

N-BOC-L-tert-Leucine, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  368010050

 View more versions of this product

Catalog No. AC368010050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 231.29
Quantity 5g
Color White to slightly yellow
Synonym N-(tert-Butoxycarbonyl)-L-leucine
Specific Rotation -4.0° to -5.0° (20°C, 589nm) (c=1, acetic acid)
Assay Percent Range 98%
Packaging Glass bottle
Chemical Name or Material N-BOC-L-tert-Leucine
CAS 62965-35-9
Molecular Formula C11H21NO4
CAS Min % 97.5
MDL Number MFCD00065574
CAS Max % 100.0
Infrared Spectrum Authentic

Specifications

Chemical Name or Material N-BOC-L-tert-Leucine
CAS 62965-35-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H21NO4
Formula Weight 231.29
MDL Number MFCD00065574
Synonym N-(tert-Butoxycarbonyl)-L-leucine