missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-serine methyl ester, 95%

$56.59 - $197.39

Chemical Identifiers

CAS 2766-43-0
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.237
MDL Number MFCD00191869
InChI Key SANNKFASHWONFD-LURJTMIESA-N
Synonym boc-ser-ome, boc-l-serine methyl ester, n-tert-butoxycarbonyl-l-serine methyl ester, n-boc-l-serine methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-l-ser-ome, methyl tert-butoxycarbonyl-l-serinate, serine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-ser-ome; boc-l-serine methyl ester
PubChem CID 7019177
IUPAC Name methyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(CO)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5988904 View Documents Alfa Aesar™
H5988904
2g Each for $56.59
 
AAH5988909 View Documents Alfa Aesar™
H5988909
10g Each for $197.39
 
Description

Description

Applications
N-tert-Butoxycarbonyl-L-serine Methyl Ester is an amino acid derivative used in the preparation of Tn antigen, L-glutamic acid p-nitroanilide analogs and new biomedical polymers having Serine and Threonine side groups.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

2766-43-0
219.237
SANNKFASHWONFD-LURJTMIESA-N
7019177
CC(C)(C)OC(=O)NC(CO)C(=O)OC
C9H17NO5
MFCD00191869
boc-ser-ome, boc-l-serine methyl ester, n-tert-butoxycarbonyl-l-serine methyl ester, n-boc-l-serine methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-l-ser-ome, methyl tert-butoxycarbonyl-l-serinate, serine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-ser-ome; boc-l-serine methyl ester
methyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate

Specifications

113°C (235°F)
2g
C9H17NO5
MFCD00191869
boc-ser-ome, boc-l-serine methyl ester, n-tert-butoxycarbonyl-l-serine methyl ester, n-boc-l-serine methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-l-ser-ome, methyl tert-butoxycarbonyl-l-serinate, serine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-ser-ome; boc-l-serine methyl ester
CC(C)(C)OC(=O)NC(CO)C(=O)OC
219.237
219.23
N-Boc-L-serine methyl ester
−18° (c=5 in Methanol)
2766-43-0
HOCH2CH[NHCO2C(CH3)3]CO2CH3
3545389
SANNKFASHWONFD-LURJTMIESA-N
methyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
7019177
95%